Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NALCN
Entrez Gene ID
259232
Gene Name
sodium leak channel, non-selective
Gene Aliases
CLIFAHDD, CanIon, IHPRF, IHPRF1, INNFD, VGCNL1, bA430M15.1
Chromosome Location
Chr.13: 101053774 - 101417206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052867.2 NP_443099.1 17 - 18 2311 79
XM_011521067.2 XP_011519369.1 17 - 18 2251 79
XM_011521068.2 XP_011519370.1 17 - 18 2234 79
XM_011521069.2 XP_011519371.1 16 - 17 2164 79
XM_017020534.1 XP_016876023.1 16 - 17 2195 79
XM_017020535.1 XP_016876024.1 16 - 17 2137 79
XM_017020536.1 XP_016876025.1 12 - 13 2063 79
XM_017020537.1 XP_016876026.1 11 - 12 1452 79
GenBank mRNA AY141972.1 - 17 - 18 2205 79

View Full Product Details