The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005228.3 | NP_005219.2 | 26 - 27 | 3411 | 71 |
GenBank mRNA | AB209442.1 | - | 26 - 27 | 3310 | 71 |
AK225422.1 | - | 24 - 25 | 3062 | 71 | |
AK290352.1 | - | 26 - 27 | 3211 | 71 | |
AK294750.1 | - | 26 - 27 | 3189 | 71 | |
AY550966.1 | - | 26 - 27 | 3211 | 71 | |
BC094761.1 | - | 25 - 26 | 3287 | 71 | |
DQ088980.1 | - | 26 - 27 | 3211 | 71 | |
GU255993.1 | - | 26 - 27 | 3208 | 71 | |
HQ912715.1 | - | 25 - 26 | 3276 | 71 | |
X00588.1 | - | 26 - 27 | 3351 | 71 |