The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005228.3 | NP_005219.2 | 27 - 28 | 3517 | 61 |
GenBank mRNA | AB209442.1 | - | 27 - 28 | 3416 | 61 |
AK225422.1 | - | 25 - 26 | 3168 | 61 | |
AK290352.1 | - | 27 - 28 | 3317 | 61 | |
AK294750.1 | - | 27 - 28 | 3295 | 61 | |
AY550966.1 | - | 27 - 28 | 3317 | 61 | |
DQ088980.1 | - | 27 - 28 | 3317 | 61 | |
HQ912715.1 | - | 26 - 27 | 3382 | 61 | |
X00588.1 | - | 27 - 28 | 3457 | 61 |