Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
Chromosome Location
Chr.7: 55019032 - 55207338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005228.3 NP_005219.2 8 - 9 1253 144
NM_201282.1 NP_958439.1 8 - 9 1253 144
NM_201283.1 NP_958440.1 8 - 9 1253 144
NM_201284.1 NP_958441.1 8 - 9 1253 144
GenBank mRNA AB209442.1 - 8 - 9 1152 144
AF125253.1 - 8 - 9 1252 144
AK290352.1 - 8 - 9 1053 144
AK294750.1 - 8 - 9 1031 144
AY550966.1 - 8 - 9 1053 144
AY698023.1 - 8 - 9 1007 144
AY698024.1 - 8 - 9 1007 144
BC094761.1 - 7 - 8 1129 144
BC118665.1 - 3 - 4 389 144
BC128419.1 - 2 - 3 192 144
DQ088980.1 - 8 - 9 1053 144
GU255993.1 - 8 - 9 1050 144
HQ912715.1 - 7 - 8 1118 144
K03193.1 - 8 - 9 1166 144
U48722.1 - 8 - 9 1251 144
U95089.1 - 9 - 10 1510 144
X00588.1 - 8 - 9 1193 144
X00663.1 - 5 - 6 581 144

View Full Product Details