The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172649.3 | NP_001166120.1 | 1 - 2 | 941 | 79 |
NM_001291106.1 | NP_001278035.1 | 1 - 2 | 941 | 79 | |
NM_001291107.1 | NP_001278036.1 | 1 - 2 | 941 | 79 | |
NM_007185.6 | NP_009116.3 | 1 - 2 | 941 | 79 | |
XM_006711139.2 | XP_006711202.1 | 2 - 3 | 169 | 79 | |
XM_006711141.2 | XP_006711204.1 | 2 - 3 | 169 | 79 | |
XM_011509111.2 | XP_011507413.1 | 1 - 2 | 789 | 79 | |
GenBank mRNA | AF329264.1 | - | 1 - 2 | 141 | 79 |
AL519587.3 | - | 2 - 3 | 173 | 79 | |
AY165003.1 | - | 1 - 2 | 931 | 79 | |
BC004145.2 | - | 2 - 3 | 119 | 79 | |
BC041088.1 | - | 1 - 2 | 173 | 79 | |
BC052491.1 | - | 1 - 2 | 915 | 79 | |
BC104758.1 | - | 1 - 2 | 204 | 79 | |
BC143226.1 | - | 1 - 2 | 204 | 79 |