The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004045.3 | NP_004036.1 | 2 - 3 | 173 | 70 |
GenBank mRNA | AK128210.1 | - | 2 - 3 | 1050 | 70 |
AK293063.1 | - | 2 - 3 | 180 | 70 | |
BC112248.1 | - | 2 - 3 | 255 | 70 | |
BC112250.1 | - | 2 - 3 | 255 | 70 | |
BI599233.1 | - | 2 - 3 | 181 | 70 | |
BT009786.1 | - | 1 - 2 | 75 | 70 | |
BU172675.1 | - | 2 - 3 | 551 | 70 | |
CN280769.1 | - | 2 - 3 | 150 | 70 | |
CR456901.1 | - | 1 - 2 | 75 | 70 |