The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171976.1 | NP_001165447.1 | 25 - 26 | 3548 | 88 |
NM_017675.4 | NP_060145.3 | 25 - 26 | 3362 | 88 | |
GenBank mRNA | AB047004.1 | - | 24 - 25 | 3312 | 88 |
AK000131.1 | - | 10 - 11 | 1501 | 88 | |
AK000390.1 | - | 20 - 21 | 2916 | 88 | |
AK225217.1 | - | 10 - 11 | 1501 | 88 | |
AK315055.1 | - | 25 - 26 | 3548 | 88 | |
BC130282.1 | - | 24 - 25 | 3321 | 88 |