The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171976.1 | NP_001165447.1 | 4 - 5 | 533 | 86 |
NM_017675.4 | NP_060145.3 | 4 - 5 | 347 | 86 | |
GenBank mRNA | AA130780.1 | - | 4 - 5 | 324 | 87 |
AB047004.1 | - | 3 - 4 | 297 | 86 | |
AK315055.1 | - | 4 - 5 | 533 | 86 | |
BC130282.1 | - | 3 - 4 | 306 | 86 | |
BP271197.1 | - | 4 - 5 | 347 | 86 | |
BX478954.1 | - | 4 - 5 | 334 | 86 |