Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD17
Entrez Gene ID
388419
Gene Name
BTB domain containing 17
Gene Aliases
BTBD17A, LGALS3BPL, TANGO10A
Chromosome Location
Chr.17: 74356416 - 74363655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011524791.2 XP_011523093.1 1 - 2 641 85
XM_017024622.1 XP_016880111.1 1 - 2 641 85

View Full Product Details