Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTY
Entrez Gene ID
7404
Gene Name
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases
KDM6AL, UTY1
Chromosome Location
Chr.Y: 13230770 - 13480670 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258249.1 NP_001245178.1 29 - 30 5319 134
NM_001258250.1 NP_001245179.1 27 - 28 4608 134
NM_001258251.1 NP_001245180.1 28 - 29 5076 134
NM_001258252.1 NP_001245181.1 28 - 29 5163 134
NM_001258255.1 NP_001245184.1 28 - 29 4791 134
NM_001258257.1 NP_001245186.1 27 - 28 4656 134
NM_001258258.1 NP_001245187.1 28 - 29 5184 134
NM_001258259.1 NP_001245188.1 27 - 28 4980 134
NM_001258260.1 NP_001245189.1 28 - 29 5118 134
NM_001258261.1 NP_001245190.1 28 - 29 5154 134
NM_001258262.1 NP_001245191.1 29 - 30 4947 134
NM_001258264.1 NP_001245193.1 28 - 29 5115 134
NM_001258265.1 NP_001245194.1 23 - 24 4779 134
NM_001258266.1 NP_001245195.1 27 - 28 5088 134
NM_007125.4 NP_009056.3 27 - 28 5028 134
NR_047596.1 26 - 27 4255 134
NR_047597.1 28 - 29 5134 134
NR_047598.1 30 - 31 5332 134
NR_047600.1 28 - 29 4952 134
NR_047602.1 26 - 27 4993 134
NR_047603.1 28 - 29 5151 134
NR_047604.1 27 - 28 4121 134
NR_047605.1 27 - 28 4312 134
NR_047606.1 27 - 28 4391 134
NR_047607.1 29 - 30 5374 134
NR_047608.1 28 - 29 4410 134
NR_047609.1 27 - 28 4786 134
NR_047610.1 27 - 28 4323 134
NR_047611.1 27 - 28 4390 134
NR_047612.1 28 - 29 4546 134
NR_047613.1 26 - 27 4308 134
NR_047614.1 29 - 30 4527 134
NR_047616.1 27 - 28 4307 134
NR_047617.1 29 - 30 4424 134
NR_047618.1 31 - 32 5129 134
NR_047621.1 28 - 29 4495 134
NR_047622.1 27 - 28 4447 134
NR_047627.1 25 - 26 4270 134
NR_047629.1 27 - 28 4522 134
NR_047630.1 25 - 26 4180 134
NR_047631.1 27 - 28 4347 134
NR_047632.1 29 - 30 4804 134
NR_047633.1 29 - 30 4304 134
NR_047634.1 26 - 27 4254 134
NR_047636.1 28 - 29 4393 134
NR_047639.1 25 - 26 4161 134
NR_047640.1 27 - 28 4239 134
NR_047641.1 28 - 29 5096 134
NR_047642.1 28 - 29 5133 134
NR_047643.1 28 - 29 4628 134
NR_047644.1 29 - 30 5221 134
NR_047645.1 28 - 29 5234 134
NR_047646.1 26 - 27 4901 134
NR_047647.1 27 - 28 4985 134
XM_011531453.2 XP_011529755.1 30 - 31 4713 134
XM_011531458.2 XP_011529760.1 29 - 30 4599 134
GenBank mRNA AF000994.1 - 27 - 28 5028 134
AK308952.1 - 12 - 13 1854 134
BC051718.1 - 7 - 8 967 134

View Full Product Details