The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002000.2 | NP_001002000.1 | 6 - 7 | 1008 | 109 |
NM_001002001.2 | NP_001002001.1 | 6 - 7 | 949 | 109 | |
NM_001002002.2 | NP_001002002.1 | 6 - 7 | 688 | 109 | |
NM_001283021.1 | NP_001269950.1 | 7 - 8 | 1084 | 109 | |
NM_001283022.1 | NP_001269951.1 | 5 - 6 | 929 | 109 | |
NM_001283023.1 | NP_001269952.1 | 5 - 6 | 604 | 109 | |
NM_016576.4 | NP_057660.2 | 5 - 6 | 929 | 109 | |
NR_104265.1 | 6 - 7 | 1032 | 109 | ||
XM_005267740.3 | XP_005267797.1 | 6 - 7 | 814 | 109 | |
XM_005267741.3 | XP_005267798.1 | 6 - 7 | 873 | 109 | |
XM_005267742.3 | XP_005267799.1 | 6 - 7 | 692 | 109 | |
XM_006720165.2 | XP_006720228.1 | 7 - 8 | 792 | 109 | |
XM_017021356.1 | XP_016876845.1 | 7 - 8 | 1008 | 109 | |
XM_017021357.1 | XP_016876846.1 | 7 - 8 | 1047 | 109 | |
XM_017021358.1 | XP_016876847.1 | 7 - 8 | 1148 | 109 | |
XM_017021359.1 | XP_016876848.1 | 7 - 8 | 791 | 109 | |
XM_017021360.1 | XP_016876849.1 | 4 - 5 | 395 | 109 | |
GenBank mRNA | AB032903.1 | - | 5 - 6 | 555 | 109 |
AF086919.1 | - | 6 - 7 | 724 | 109 | |
AF135159.1 | - | 6 - 7 | 894 | 109 | |
AF419346.1 | - | 6 - 7 | 798 | 109 | |
AK226166.1 | - | 7 - 8 | 840 | 109 | |
BC003053.1 | - | 5 - 6 | 878 | 109 | |
BC008021.1 | - | 5 - 6 | 877 | 109 | |
BC009832.2 | - | 6 - 7 | 606 | 109 | |
BC093039.1 | - | 6 - 7 | 747 | 109 | |
BX161436.1 | - | 6 - 7 | 929 | 109 | |
BX161518.1 | - | 6 - 7 | 819 | 109 | |
BX247993.1 | - | 5 - 6 | 527 | 109 | |
CR457156.1 | - | 5 - 6 | 547 | 109 |