The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002002.2 | NP_001002002.1 | 1 - 2 | 107 | 87 |
NM_001283023.1 | NP_001269952.1 | 1 - 2 | 107 | 87 | |
XM_005267742.3 | XP_005267799.1 | 1 - 2 | 111 | 87 | |
XM_006720165.2 | XP_006720228.1 | 1 - 2 | 76 | 87 | |
XM_017021359.1 | XP_016876848.1 | 1 - 2 | 75 | 87 | |
GenBank mRNA | AV660418.1 | - | 1 - 2 | 86 | 87 |
BP347823.1 | - | 1 - 2 | 107 | 87 |