Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
Chromosome Location
Chr.2: 121337776 - 121649476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142273.1 NP_001135745.1 10 - 11 1336 136
NM_001142274.1 NP_001135746.1 10 - 11 1336 136
NM_001207051.1 NP_001193980.1 10 - 11 1336 136
NM_015282.2 NP_056097.1 10 - 11 1336 136
XM_006712377.1 XP_006712440.1 10 - 11 1307 136
XM_006712381.1 XP_006712444.1 10 - 11 1307 136
XM_006712382.1 XP_006712445.1 10 - 11 1307 136
XM_006712383.1 XP_006712446.1 10 - 11 1307 136
XM_006712385.1 XP_006712448.1 4 - 5 379 136
XM_011510848.1 XP_011509150.1 10 - 11 1307 136
XM_011510849.1 XP_011509151.1 10 - 11 1307 136
XM_017003657.1 XP_016859146.1 10 - 11 1307 136
XM_017003658.1 XP_016859147.1 10 - 11 1307 136
XM_017003659.1 XP_016859148.1 10 - 11 1307 136
XM_017003660.1 XP_016859149.1 10 - 11 1307 136
XM_017003661.1 XP_016859150.1 10 - 11 1307 136
XM_017003662.1 XP_016859151.1 10 - 11 1307 136
XM_017003663.1 XP_016859152.1 10 - 11 1307 136
XM_017003664.1 XP_016859153.1 10 - 11 1307 136
XM_017003665.1 XP_016859154.1 10 - 11 1307 136
XM_017003666.1 XP_016859155.1 10 - 11 1307 136
XM_017003667.1 XP_016859156.1 10 - 11 1307 136
XM_017003668.1 XP_016859157.1 10 - 11 1307 136
XM_017003669.1 XP_016859158.1 10 - 11 1307 136
XM_017003670.1 XP_016859159.1 10 - 11 1307 136
XM_017003671.1 XP_016859160.1 10 - 11 1307 136
XM_017003672.1 XP_016859161.1 10 - 11 1307 136
XM_017003673.1 XP_016859162.1 10 - 11 1307 136
XM_017003674.1 XP_016859163.1 10 - 11 1307 136
XM_017003675.1 XP_016859164.1 10 - 11 1307 136
XM_017003676.1 XP_016859165.1 3 - 4 2141 136
XM_017003677.1 XP_016859166.1 3 - 4 2141 136
XM_017003678.1 XP_016859167.1 3 - 4 2140 136
XM_017003679.1 XP_016859168.1 3 - 4 2141 136
XM_017003680.1 XP_016859169.1 3 - 4 2141 136
XM_017003681.1 XP_016859170.1 3 - 4 2140 136
XM_017003682.1 XP_016859171.1 3 - 4 2140 136
XM_017003683.1 XP_016859172.1 3 - 4 2139 136
XM_017003684.1 XP_016859173.1 3 - 4 1122 136
XM_017003685.1 XP_016859174.1 3 - 4 2139 136
XM_017003686.1 XP_016859175.1 3 - 4 2139 136
XM_017003687.1 XP_016859176.1 3 - 4 1122 136
XM_017003688.1 XP_016859177.1 3 - 4 2139 136
XM_017003689.1 XP_016859178.1 3 - 4 1121 136
XM_017003690.1 XP_016859179.1 10 - 11 1307 136
XM_017003691.1 XP_016859180.1 10 - 11 1307 136
GenBank mRNA AB014522.1 - 2 - 3 199 136
AF347693.1 - 9 - 10 1111 136
AK091140.1 - 10 - 11 1297 136
AK297259.1 - 3 - 4 290 136
AK299331.1 - 5 - 6 858 136
AK302393.1 - 4 - 5 396 136
BC112940.1 - 10 - 11 1280 136
BC132723.1 - 10 - 11 1298 136
BC144107.1 - 10 - 11 1298 136
CR933722.1 - 3 - 4 288 136

View Full Product Details