Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
Chromosome Location
Chr.2: 121337776 - 121649476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142273.1 NP_001135745.1 4 - 5 770 103
NM_001142274.1 NP_001135746.1 4 - 5 770 103
NM_001207051.1 NP_001193980.1 4 - 5 770 103
NM_015282.2 NP_056097.1 4 - 5 770 103
XM_006712377.1 XP_006712440.1 4 - 5 741 103
XM_006712381.1 XP_006712444.1 4 - 5 741 103
XM_006712382.1 XP_006712445.1 4 - 5 741 103
XM_006712383.1 XP_006712446.1 4 - 5 741 103
XM_011510848.1 XP_011509150.1 4 - 5 741 103
XM_011510849.1 XP_011509151.1 4 - 5 741 103
XM_017003657.1 XP_016859146.1 4 - 5 741 103
XM_017003658.1 XP_016859147.1 4 - 5 741 103
XM_017003659.1 XP_016859148.1 4 - 5 741 103
XM_017003660.1 XP_016859149.1 4 - 5 741 103
XM_017003661.1 XP_016859150.1 4 - 5 741 103
XM_017003662.1 XP_016859151.1 4 - 5 741 103
XM_017003663.1 XP_016859152.1 4 - 5 741 103
XM_017003664.1 XP_016859153.1 4 - 5 741 103
XM_017003665.1 XP_016859154.1 4 - 5 741 103
XM_017003666.1 XP_016859155.1 4 - 5 741 103
XM_017003667.1 XP_016859156.1 4 - 5 741 103
XM_017003668.1 XP_016859157.1 4 - 5 741 103
XM_017003669.1 XP_016859158.1 4 - 5 741 103
XM_017003670.1 XP_016859159.1 4 - 5 741 103
XM_017003671.1 XP_016859160.1 4 - 5 741 103
XM_017003672.1 XP_016859161.1 4 - 5 741 103
XM_017003673.1 XP_016859162.1 4 - 5 741 103
XM_017003674.1 XP_016859163.1 4 - 5 741 103
XM_017003675.1 XP_016859164.1 4 - 5 741 103
XM_017003690.1 XP_016859179.1 4 - 5 741 103
XM_017003691.1 XP_016859180.1 4 - 5 741 103
GenBank mRNA AF347693.1 - 3 - 4 545 103
AJ288057.1 - 4 - 5 770 103
AK091140.1 - 4 - 5 731 103
BC112940.1 - 4 - 5 714 103
BC132723.1 - 4 - 5 732 103
BC144107.1 - 4 - 5 732 103

View Full Product Details