Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLII
Entrez Gene ID
2314
Gene Name
FLII, actin remodeling protein
Gene Aliases
FLI, FLIL, Fli1
Chromosome Location
Chr.17: 18244815 - 18258916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256264.1 NP_001243193.1 13 - 14 1640 56
NM_001256265.1 NP_001243194.1 12 - 13 1654 56
NM_002018.3 NP_002009.1 13 - 14 1819 56
XM_005256555.3 XP_005256612.1 13 - 14 1608 56
XM_005256556.3 XP_005256613.1 13 - 14 1605 56
XM_005256558.2 XP_005256615.1 13 - 14 1605 56
GenBank mRNA AB208864.1 - 13 - 14 1554 56
AK295655.1 - 12 - 13 1461 56
AK295814.1 - 13 - 14 1722 56
AK297836.1 - 13 - 14 1553 56
AK310400.1 - 13 - 14 1641 56
BC021885.1 - 11 - 12 1384 56
BC025300.1 - 13 - 14 1644 56
U01184.1 - 13 - 14 1591 56

View Full Product Details