Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD3
Entrez Gene ID
29766
Gene Name
tropomodulin 3
Gene Aliases
UTMOD
Chromosome Location
Chr.15: 51829628 - 51912138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014547.4 NP_055362.1 7 - 8 1014 89
XM_017022087.1 XP_016877576.1 8 - 9 1045 89
XM_017022088.1 XP_016877577.1 7 - 8 886 89
GenBank mRNA AF177171.1 - 7 - 8 976 89
AF237631.1 - 6 - 7 796 89
AK310344.1 - 5 - 6 693 89
AK312569.1 - 7 - 8 1011 89
BC020542.2 - 7 - 8 950 89
BX647277.1 - 7 - 8 976 89

View Full Product Details