The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193455.1 | NP_001180384.1 | 13 - 14 | 1876 | 159 |
NM_017755.5 | NP_060225.4 | 14 - 15 | 1981 | 159 | |
NR_037947.1 | 13 - 14 | 1896 | 159 | ||
GenBank mRNA | AB255451.1 | - | - | 1980 | 159 |
AK000310.1 | - | 8 - 9 | 977 | 159 | |
AK023994.1 | - | 2 - 3 | 247 | 159 | |
AK055456.1 | - | 13 - 14 | 1826 | 159 | |
AK291144.1 | - | 14 - 15 | 1664 | 159 | |
AK298980.1 | - | 13 - 14 | 1558 | 159 | |
BC001041.2 | - | 12 - 13 | 1337 | 159 | |
BC137083.1 | - | 14 - 15 | 1662 | 159 |