The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193455.1 | NP_001180384.1 | 5 - 6 | 897 | 96 |
NM_017755.5 | NP_060225.4 | 6 - 7 | 1002 | 96 | |
GenBank mRNA | AB255451.1 | - | - | 1001 | 96 |
AK291144.1 | - | 6 - 7 | 685 | 96 | |
AK298980.1 | - | 5 - 6 | 579 | 96 | |
BC001041.2 | - | 4 - 5 | 358 | 96 | |
BC137083.1 | - | 6 - 7 | 683 | 96 | |
BG387061.1 | - | 5 - 6 | 573 | 96 | |
BQ689094.1 | - | 6 - 7 | 644 | 96 |