Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARRDC1
Entrez Gene ID
92714
Gene Name
arrestin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 137605640 - 137615360 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317968.1 NP_001304897.1 2 - 3 308 96
NM_152285.3 NP_689498.1 2 - 3 308 96
XM_005266119.1 XP_005266176.1 2 - 3 307 96
XM_006717320.3 XP_006717383.1 1 - 2 679 96
XM_006717321.3 XP_006717384.1 1 - 2 679 96
XM_006717322.3 XP_006717385.1 1 - 2 679 96
XM_017015288.1 XP_016870777.1 1 - 2 679 96
GenBank mRNA AJ420420.1 - 2 - 3 271 96
AK001822.1 - 1 - 2 568 96
AK308376.1 - 2 - 3 245 96
AL542109.3 - 2 - 3 304 96
BC032346.1 - 2 - 3 294 96
BI906381.1 - 2 - 3 309 96

View Full Product Details