The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271878.1 | NP_001258807.1 | 4 - 5 | 1090 | 136 |
NM_006559.2 | NP_006550.1 | 5 - 6 | 1207 | 136 | |
NR_073498.1 | 5 - 6 | 1207 | 136 | ||
NR_073499.1 | 5 - 6 | 1207 | 136 | ||
GenBank mRNA | AK091346.1 | - | 6 - 7 | 960 | 136 |
AK293369.1 | - | 6 - 7 | 988 | 136 | |
AK303218.1 | - | 6 - 7 | 952 | 136 | |
BC000717.1 | - | 5 - 6 | 1019 | 136 | |
BC010132.2 | - | 5 - 6 | 975 | 136 | |
BC019109.2 | - | 5 - 6 | 1019 | 136 | |
M88108.1 | - | 5 - 6 | 1011 | 136 | |
U78971.1 | - | 4 - 5 | 788 | 136 |