The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015630.3 | NP_056445.3 | 10 - 11 | 1750 | 95 |
XM_011510943.2 | XP_011509245.1 | 9 - 10 | 1634 | 95 | |
GenBank mRNA | AF286904.1 | - | 10 - 11 | 1742 | 95 |
AK001433.1 | - | 8 - 9 | 1341 | 95 | |
AK002010.1 | - | 1 - 2 | 197 | 95 | |
AK125789.1 | - | 11 - 12 | 1819 | 95 | |
BC093818.1 | - | 10 - 11 | 1662 | 95 | |
BC093820.1 | - | 10 - 11 | 1662 | 95 |