The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015630.3 | NP_056445.3 | 11 - 12 | 1883 | 80 |
XM_011510941.2 | XP_011509243.1 | 11 - 12 | 2088 | 80 | |
XM_011510943.2 | XP_011509245.1 | 10 - 11 | 1767 | 80 | |
GenBank mRNA | AF286904.1 | - | 11 - 12 | 1875 | 80 |
AK001433.1 | - | 9 - 10 | 1474 | 80 | |
AK002010.1 | - | 2 - 3 | 330 | 80 | |
AK125789.1 | - | 12 - 13 | 1952 | 80 | |
BC093818.1 | - | 11 - 12 | 1795 | 80 | |
BC093820.1 | - | 11 - 12 | 1795 | 80 |