The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019592.6 | NP_062538.5 | 11 - 12 | 1501 | 76 |
XM_011518862.1 | XP_011517164.1 | 11 - 12 | 1778 | 76 | |
GenBank mRNA | AF265230.1 | - | 11 - 12 | 1499 | 76 |
AK000389.1 | - | 10 - 11 | 1392 | 76 | |
AK000697.1 | - | 10 - 11 | 1392 | 76 | |
AK022300.1 | - | 6 - 7 | 773 | 76 | |
AK022532.1 | - | 11 - 12 | 1497 | 76 | |
AK314401.1 | - | 11 - 12 | 1456 | 76 | |
AL834272.1 | - | 1 - 2 | 53 | 76 | |
BC016691.1 | - | 12 - 13 | 1456 | 76 | |
BC032618.1 | - | 7 - 8 | 824 | 76 | |
BC063115.1 | - | 11 - 12 | 1482 | 76 | |
BC110584.1 | - | 11 - 12 | 1494 | 76 | |
BC110585.1 | - | 11 - 12 | 1494 | 76 | |
BC152309.1 | - | 11 - 12 | 1494 | 76 |