The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019592.6 | NP_062538.5 | 2 - 3 | 219 | 97 |
XM_011518862.1 | XP_011517164.1 | 2 - 3 | 496 | 97 | |
GenBank mRNA | AF265230.1 | - | 2 - 3 | 217 | 97 |
AK000389.1 | - | 1 - 2 | 110 | 97 | |
AK000697.1 | - | 1 - 2 | 110 | 97 | |
AK314401.1 | - | 2 - 3 | 174 | 97 | |
BC016691.1 | - | 3 - 4 | 174 | 97 | |
BC063115.1 | - | 2 - 3 | 200 | 97 | |
BC110584.1 | - | 2 - 3 | 212 | 97 | |
BC110585.1 | - | 2 - 3 | 212 | 97 | |
BC152309.1 | - | 2 - 3 | 212 | 97 | |
DB273543.1 | - | 2 - 3 | 219 | 97 |