Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr.9: 100302075 - 100352977 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161584.1 NP_001155056.1 7 - 8 1779 114
NM_017746.3 NP_060216.2 7 - 8 1835 114
XM_017014848.1 XP_016870337.1 7 - 8 1324 114
GenBank mRNA AB060968.1 - 7 - 8 1751 114
AJ420436.1 - 2 - 3 260 114
AK000294.1 - 7 - 8 1751 114
AK298916.1 - 9 - 10 1358 114
AK302619.1 - 7 - 8 1782 114
AY598337.1 - 6 - 7 1620 114
BC030652.2 - 7 - 8 1671 114

View Full Product Details