The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001688.4 | NP_001679.2 | 6 - 7 | 1113 | 61 |
GenBank mRNA | AK223018.1 | - | 6 - 7 | 738 | 61 |
AK291077.1 | - | 6 - 7 | 770 | 61 | |
AK311287.1 | - | 2 - 3 | 150 | 61 | |
AK313541.1 | - | 6 - 7 | 740 | 61 | |
AY871272.1 | - | 6 - 7 | 747 | 61 | |
BC005366.1 | - | 6 - 7 | 758 | 61 | |
BC005960.1 | - | 6 - 7 | 750 | 61 | |
BC016350.1 | - | 6 - 7 | 789 | 61 | |
X60221.1 | - | 6 - 7 | 724 | 61 |