The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024025.2 | NP_076930.1 | 1 - 2 | 441 | 122 |
GenBank mRNA | AB103376.1 | - | 1 - 2 | 256 | 122 |
AB158288.1 | - | 1 - 2 | 318 | 122 | |
AB237597.1 | - | 1 - 2 | 253 | 122 | |
AY902194.1 | - | 1 - 2 | 256 | 122 | |
BC001613.1 | - | 1 - 2 | 256 | 122 |