Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
Chromosome Location
Chr.19: 7688757 - 7702755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207019.2 NP_001193948.2 8 - 9 703 60
NM_001220500.1 NP_001207429.1 9 - 10 839 60
NM_002002.4 NP_001993.2 9 - 10 835 60
XM_005272462.4 XP_005272519.1 9 - 10 1440 60
GenBank mRNA AK223207.1 - 9 - 10 804 60
AK310071.1 - 8 - 9 984 60
BC014108.2 - 9 - 10 800 60
BC062591.1 - 9 - 10 793 60
BC064417.1 - 8 - 9 1117 60
M14766.1 - 9 - 10 807 60
M15059.1 - 9 - 10 835 60
X04772.1 - 9 - 10 835 60

View Full Product Details