Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP85
Entrez Gene ID
64793
Gene Name
centrosomal protein 85
Gene Aliases
CCDC21
Chromosome Location
Chr.1: 26234153 - 26278810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281517.2 NP_001268446.1 12 - 13 2157 78
NM_001281518.2 NP_001268447.1 6 - 7 718 78
NM_022778.4 NP_073615.2 13 - 14 2310 78
GenBank mRNA AK025653.1 - 7 - 8 909 78
AK299312.1 - 12 - 13 2110 78
AL137420.1 - 7 - 8 907 78
BC019902.1 - 6 - 7 711 78
BC064528.1 - 13 - 14 2261 78

View Full Product Details