Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCRN3
Entrez Gene ID
79634
Gene Name
secernin 3
Gene Aliases
SES3
Chromosome Location
Chr.2: 174395729 - 174430071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193528.1 NP_001180457.1 7 - 8 1099 80
NM_024583.4 NP_078859.2 7 - 8 1190 80
XM_005246853.2 XP_005246910.1 7 - 8 1595 80
XM_005246854.4 XP_005246911.1 7 - 8 2755 80
XM_005246855.2 XP_005246912.1 7 - 8 1643 80
XM_005246856.2 XP_005246913.1 7 - 8 1599 80
XM_005246857.2 XP_005246914.1 7 - 8 1299 80
XM_011511839.2 XP_011510141.1 6 - 7 999 80
XM_017004909.1 XP_016860398.1 7 - 8 2187 80
XM_017004910.1 XP_016860399.1 7 - 8 1665 80
GenBank mRNA AF279776.1 - 3 - 4 527 80
AK026795.1 - 3 - 4 537 80
AK057912.1 - 7 - 8 1168 80
AK295357.1 - 7 - 8 1083 80
AK296558.1 - 6 - 7 989 80
BC031821.1 - 4 - 5 651 80
BC119684.1 - 7 - 8 1141 80
BC119685.1 - 7 - 8 1141 80

View Full Product Details