The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005408.1 | NP_001005408.1 | 1 - 2 | 292 | 65 |
NM_032442.2 | NP_115818.2 | 1 - 2 | 292 | 65 | |
GenBank mRNA | AB058690.1 | - | 1 - 2 | 292 | 65 |
AL136870.1 | - | 1 - 2 | 148 | 65 |