The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005408.1 | NP_001005408.1 | 5 - 6 | 1212 | 55 |
NM_032442.2 | NP_115818.2 | 5 - 6 | 1212 | 55 | |
GenBank mRNA | AB058690.1 | - | 5 - 6 | 1212 | 55 |
AL136870.1 | - | 5 - 6 | 1068 | 55 | |
AL833570.1 | - | 5 - 6 | 458 | 55 |