Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
Chromosome Location
Chr.6: 34246380 - 34249108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008703.2 NP_001008703.1 3 - 4 345 95
NM_178508.4 NP_848603.2 3 - 4 597 95
GenBank mRNA AK290164.1 - 3 - 4 279 95
BC047919.1 - 3 - 4 478 95
BU956518.1 - 2 - 3 84 95
CK001844.1 - 3 - 4 348 95

View Full Product Details