Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
Chromosome Location
Chr.7: 28995630 - 29146537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK124472.1 - 5 - 6 643 137

View Full Product Details