Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
Chromosome Location
Chr.7: 28995630 - 29146537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019029.2 NP_061902.2 1 - 2 198 127
GenBank mRNA BC016838.2 - 1 - 2 158 127

View Full Product Details