The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134442.4 | NP_604391.1 | 4 - 5 | 550 | 107 |
NR_135473.1 | 4 - 5 | 550 | 107 | ||
XM_011510645.1 | XP_011508947.1 | 4 - 5 | 364 | 107 | |
XM_011510646.2 | XP_011508948.1 | 4 - 5 | 526 | 107 | |
XM_011510648.2 | XP_011508950.1 | 4 - 5 | 526 | 107 | |
XM_011510649.2 | XP_011508951.1 | 3 - 4 | 361 | 107 | |
XM_011510650.2 | XP_011508952.1 | 3 - 4 | 460 | 107 | |
XM_017003401.1 | XP_016858890.1 | 2 - 3 | 159 | 107 | |
GenBank mRNA | AK302333.1 | - | 3 - 4 | 319 | 107 |
AK312747.1 | - | 5 - 6 | 445 | 107 | |
AY347527.1 | - | 4 - 5 | 342 | 107 | |
BC010636.1 | - | 4 - 5 | 393 | 107 | |
BT019509.1 | - | 3 - 4 | 299 | 107 | |
M34356.1 | - | 3 - 4 | 299 | 107 | |
X55545.1 | - | - | 442 | 107 |