Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATSL3
Entrez Gene ID
652968
Gene Name
GATS protein like 3
Gene Aliases
CASTOR1
Chromosome Location
Chr.22: 30285118 - 30289627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037666.2 NP_001032755.1 4 - 5 634 70
GenBank mRNA AK055587.1 - 4 - 5 540 70
AK309436.1 - 3 - 4 467 70
BC021927.1 - 4 - 5 520 70
CR456449.1 - 4 - 5 549 70

View Full Product Details