Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCB1
Entrez Gene ID
9657
Gene Name
IQ motif containing B1
Gene Aliases
NPHP5, PIQ, SLSN5
Chromosome Location
Chr.3: 121769761 - 121835079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001023570.3 NP_001018864.2 2 - 3 199 95
NM_001023571.3 NP_001018865.2 2 - 3 199 95
XM_005247911.3 XP_005247968.1 2 - 3 198 95
XM_005247912.2 XP_005247969.1 2 - 3 175 95
XM_011513335.2 XP_011511637.1 2 - 3 175 95
XM_017007537.1 XP_016863026.1 2 - 3 175 95
XM_017007539.1 XP_016863028.1 2 - 3 185 95
GenBank mRNA AY714228.1 - 2 - 3 193 95
AY964667.1 - 2 - 3 149 95
BC005806.2 - 2 - 3 140 95
BG721354.1 - - 204 95
BQ068103.1 - - 185 95
D25278.1 - 2 - 3 140 95

View Full Product Details