The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001023570.3 | NP_001018864.2 | 12 - 13 | 1491 | 118 |
NM_001023571.3 | NP_001018865.2 | 9 - 10 | 1092 | 118 | |
NM_001319107.1 | NP_001306036.1 | 12 - 13 | 1490 | 118 | |
NR_134968.1 | 11 - 12 | 1380 | 118 | ||
XM_005247911.3 | XP_005247968.1 | 12 - 13 | 1490 | 118 | |
XM_005247912.2 | XP_005247969.1 | 11 - 12 | 1304 | 118 | |
XM_011513335.2 | XP_011511637.1 | 10 - 11 | 1210 | 118 | |
XM_017007537.1 | XP_016863026.1 | 11 - 12 | 1373 | 118 | |
XM_017007538.1 | XP_016863027.1 | 8 - 9 | 1175 | 118 | |
XM_017007539.1 | XP_016863028.1 | 9 - 10 | 1078 | 118 | |
GenBank mRNA | AB062481.1 | - | 11 - 12 | 1374 | 118 |
AY714228.1 | - | 12 - 13 | 1485 | 118 | |
AY964667.1 | - | 9 - 10 | 1042 | 118 | |
AY964668.1 | - | 12 - 13 | 1429 | 118 | |
BC005806.2 | - | 9 - 10 | 1033 | 118 | |
D25278.1 | - | 12 - 13 | 1432 | 118 |