The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000521.3 | NP_000512.1 | 12 - 13 | 1618 | 95 |
NM_001292004.1 | NP_001278933.1 | 12 - 13 | 1738 | 95 | |
GenBank mRNA | AF378118.1 | - | 12 - 13 | 1573 | 95 |
AK122992.1 | - | 13 - 14 | 1475 | 95 | |
AK130002.1 | - | 12 - 13 | 1602 | 95 | |
AK130375.1 | - | 12 - 13 | 1529 | 95 | |
AY643499.1 | - | 12 - 13 | 1743 | 95 | |
BC017378.2 | - | 12 - 13 | 1523 | 95 | |
BT009919.1 | - | 12 - 13 | 1501 | 95 | |
FJ224336.1 | - | 12 - 13 | 1743 | 95 | |
GQ472222.1 | - | 12 - 13 | 1743 | 95 | |
M13519.1 | - | 12 - 13 | 1549 | 95 | |
M19735.1 | - | 12 - 13 | 1465 | 95 |