The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130447.1 | NP_001123919.1 | 4 - 5 | 470 | 137 |
NM_001323630.1 | NP_001310559.1 | 4 - 5 | 430 | 137 | |
NM_001323631.1 | NP_001310560.1 | 6 - 7 | 652 | 137 | |
NM_001323632.1 | NP_001310561.1 | 4 - 5 | 493 | 137 | |
NM_018097.2 | NP_060567.1 | 5 - 6 | 563 | 137 | |
XM_017022396.1 | XP_016877885.1 | 5 - 6 | 567 | 137 | |
XM_017022397.1 | XP_016877886.1 | 5 - 6 | 534 | 137 | |
GenBank mRNA | AK001322.1 | - | 5 - 6 | 521 | 137 |
AK022939.1 | - | 5 - 6 | 534 | 137 | |
BC010903.1 | - | 5 - 6 | 542 | 137 | |
DA923763.1 | - | 4 - 5 | 427 | 137 |