The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171.5 | NP_001162.4 | 13 - 14 | 1813 | 93 |
XM_011522479.2 | XP_011520781.1 | 13 - 14 | 1957 | 93 | |
XM_011522480.1 | XP_011520782.1 | 13 - 14 | 1761 | 93 | |
XM_011522481.2 | XP_011520783.1 | 13 - 14 | 1960 | 93 | |
XM_011522482.2 | XP_011520784.1 | 13 - 14 | 1954 | 93 | |
XM_017023212.1 | XP_016878701.1 | 13 - 14 | 1957 | 93 | |
XM_017023213.1 | XP_016878702.1 | 13 - 14 | 1847 | 93 | |
XM_017023214.1 | XP_016878703.1 | 13 - 14 | 1958 | 93 | |
GenBank mRNA | AF076622.1 | - | 13 - 14 | 1799 | 93 |
AF168791.1 | - | 13 - 14 | 1776 | 93 | |
AM711638.1 | - | 13 - 14 | 1776 | 93 | |
AM774324.1 | - | 13 - 14 | 1776 | 93 | |
BC131732.1 | - | 13 - 14 | 1799 | 93 |