The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032336.2 | NP_115712.1 | 5 - 6 | 607 | 105 |
XM_005273659.3 | XP_005273716.1 | 5 - 6 | 516 | 105 | |
GenBank mRNA | AK095334.1 | - | 5 - 6 | 564 | 105 |
AK313373.1 | - | 5 - 6 | 552 | 105 | |
BC005995.1 | - | 5 - 6 | 586 | 105 |