The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323971.1 | NP_001310900.1 | 8 - 9 | 1567 | 65 |
NM_001323972.1 | NP_001310901.1 | 7 - 8 | 1264 | 65 | |
NM_001323973.1 | NP_001310902.1 | 8 - 9 | 1567 | 65 | |
NM_001323974.1 | NP_001310903.1 | 8 - 9 | 1481 | 65 | |
NM_001323975.1 | NP_001310904.1 | 8 - 9 | 1481 | 65 | |
NM_004594.2 | NP_004585.1 | 8 - 9 | 1487 | 65 | |
NR_136664.1 | 8 - 9 | 1481 | 65 | ||
NR_136667.1 | 8 - 9 | 1481 | 65 | ||
XM_017023594.1 | XP_016879083.1 | 8 - 9 | 2890 | 65 | |
GenBank mRNA | AB209203.1 | - | 7 - 8 | 2292 | 65 |
AF111173.1 | - | 8 - 9 | 1493 | 65 | |
BC142671.1 | - | 8 - 9 | 1450 | 65 | |
BC150207.1 | - | 7 - 8 | 1743 | 65 |