Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
Chromosome Location
Chr.16: 67229389 - 67247522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318202.1 NP_001305131.1 15 - 16 2239 121
NM_013241.2 NP_037373.2 13 - 14 2161 121
XM_006721180.1 XP_006721243.1 4 - 5 814 121
XM_011523043.2 XP_011521345.1 15 - 16 2455 121
XM_011523044.1 XP_011521346.1 15 - 16 2184 121
XM_011523045.2 XP_011521347.1 15 - 16 1949 121
GenBank mRNA AB041046.1 - 13 - 14 2131 121
AB209584.1 - 11 - 12 2630 121
AK301157.1 - 3 - 4 872 121
AY192154.1 - 13 - 14 2049 121
BC033084.1 - 13 - 14 2106 121
KJ658417.1 - 15 - 16 2127 121

View Full Product Details