The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 3 - 4 | 484 | 106 |
NM_013241.2 | NP_037373.2 | 3 - 4 | 484 | 106 | |
XM_011523043.2 | XP_011521345.1 | 3 - 4 | 700 | 106 | |
XM_011523044.1 | XP_011521346.1 | 3 - 4 | 429 | 106 | |
XM_011523045.2 | XP_011521347.1 | 3 - 4 | 194 | 106 | |
GenBank mRNA | AB041046.1 | - | 3 - 4 | 454 | 106 |
AF113615.1 | - | 3 - 4 | 390 | 106 | |
AY192154.1 | - | 3 - 4 | 372 | 106 | |
BC033084.1 | - | 3 - 4 | 429 | 106 | |
KJ658417.1 | - | 3 - 4 | 372 | 106 |