The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003869.5 | NP_003860.2 | 8 - 9 | 2331 | 93 |
NM_198061.2 | NP_932327.1 | 8 - 9 | 2331 | 93 | |
NR_036684.1 | 8 - 9 | 2569 | 93 | ||
XM_011523421.2 | XP_011521723.1 | 8 - 9 | 1769 | 93 | |
XM_017023818.1 | XP_016879307.1 | 8 - 9 | 1769 | 93 | |
GenBank mRNA | AK095522.1 | - | 8 - 9 | 2569 | 93 |
AK222724.1 | - | 8 - 9 | 1209 | 93 | |
AK290482.1 | - | 8 - 9 | 1611 | 93 | |
AL713761.1 | - | 8 - 9 | 2284 | 93 | |
BC032095.2 | - | 8 - 9 | 2315 | 93 | |
BC071874.1 | - | 8 - 9 | 1270 | 93 | |
BC098417.1 | - | 8 - 9 | 1273 | 93 | |
D50579.1 | - | 8 - 9 | 1206 | 93 | |
U60553.1 | - | 8 - 9 | 1112 | 93 | |
Y09616.1 | - | 8 - 9 | 1217 | 93 |