The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133445.2 | NP_597702.2 | 6 - 7 | 3362 | 61 |
GenBank mRNA | AB075853.1 | - | 6 - 7 | 3362 | 61 |
AF416558.1 | - | 6 - 7 | 2774 | 61 | |
AJ416950.2 | - | 6 - 7 | 2776 | 61 | |
AK127342.1 | - | 6 - 7 | 2329 | 61 | |
BC132866.1 | - | 6 - 7 | 3188 | 61 | |
BC132868.1 | - | 6 - 7 | 3188 | 61 | |
BC167432.1 | - | 6 - 7 | 3362 | 61 |