The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321881.1 | NP_001308810.1 | 13 - 14 | 1250 | 86 |
NM_001321882.1 | NP_001308811.1 | 13 - 14 | 1468 | 86 | |
NM_024635.3 | NP_078911.3 | 13 - 14 | 1254 | 86 | |
XM_005252127.3 | XP_005252184.1 | 13 - 14 | 1325 | 86 | |
XM_011518903.2 | XP_011517205.1 | 10 - 11 | 1057 | 86 | |
XM_011518904.2 | XP_011517206.1 | 7 - 8 | 743 | 86 | |
XM_017015012.1 | XP_016870501.1 | 14 - 15 | 1313 | 86 | |
GenBank mRNA | AK025266.1 | - | 13 - 14 | 1250 | 86 |
AK225237.1 | - | 13 - 14 | 1250 | 86 | |
AK292239.1 | - | 13 - 14 | 1254 | 86 | |
BC117427.1 | - | 13 - 14 | 1175 | 86 | |
BC117429.1 | - | 13 - 14 | 1171 | 86 |