The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321881.1 | NP_001308810.1 | 21 - 22 | 2170 | 124 |
NM_001321882.1 | NP_001308811.1 | 21 - 22 | 2388 | 124 | |
NM_024635.3 | NP_078911.3 | 21 - 22 | 2174 | 124 | |
XM_005252127.3 | XP_005252184.1 | 21 - 22 | 2245 | 124 | |
XM_011518903.2 | XP_011517205.1 | 18 - 19 | 1977 | 124 | |
XM_011518904.2 | XP_011517206.1 | 15 - 16 | 1663 | 124 | |
XM_017015012.1 | XP_016870501.1 | 22 - 23 | 2233 | 124 | |
GenBank mRNA | AK026296.1 | - | 5 - 6 | 509 | 124 |
AK292239.1 | - | 21 - 22 | 2174 | 124 | |
BC117427.1 | - | 21 - 22 | 2095 | 124 | |
BC117429.1 | - | 21 - 22 | 2091 | 124 |